missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichlorobenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1011830

 View more versions of this product

Catalog No. AAA1011830



Specifications

Specifications

2,3-Dichlorobenzaldehyde
143°C to 145°C (30mmHg)
250g
C7H4Cl2O
UN1759
benzaldehyde, 2,3-dichloro, dichlorobenzaldehyde, benzaldehyde, dichloro, ccris 8612, dichlorbenzaldehyd, pubchem3079, 2,3-dichlorobenzaldehyd, benzaldehyde,3-dichloro, 2,3-dichloro-benzaldehyde, acmc-209nf0
C1=CC(=C(C(=C1)Cl)Cl)C=O
175.008
175.01
135°C (275°F)
61°C to 65°C
6334-18-5
MFCD00010127
508198
LLMLNAVBOAMOEE-UHFFFAOYSA-N
2,3-dichlorobenzaldehyde
35745
98%

Chemical Identifiers

6334-18-5
175.008
LLMLNAVBOAMOEE-UHFFFAOYSA-N
35745
C1=CC(=C(C(=C1)Cl)Cl)C=O
C7H4Cl2O
MFCD00010127
benzaldehyde, 2,3-dichloro, dichlorobenzaldehyde, benzaldehyde, dichloro, ccris 8612, dichlorbenzaldehyd, pubchem3079, 2,3-dichlorobenzaldehyd, benzaldehyde,3-dichloro, 2,3-dichloro-benzaldehyde, acmc-209nf0
2,3-dichlorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : TSCA

SDS Documents