Please login to your online account to display your discounted pricing

2,3-Dichloroanisole, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC168900250

 View more versions of this product

Catalog No. AC168900250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Low Melting Solid
Assay ≥95%
Formula Weight 177.03
Color White to brownish
Quantity 25g
EINECS Number 217-853-1
TSCA TSCA
Melting Point 30° to 35°C
Beilstein 06, I, 102
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Dichloroanisole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 96%
Assay Percent Range 96%
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
MDL Number MFCD00000545
CAS 1984-59-4
Synonym 1, 2-Dichloro-3-methoxybenzene
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 3-Dichloroanisole
Name Note 96%
CAS 1984-59-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
Formula Weight 177.03
MDL Number MFCD00000545
Synonym 1, 2-Dichloro-3-methoxybenzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents