missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichloroaniline 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113095000

 View more versions of this product

Catalog No. AC113095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 3-Dichloroaniline
Authentic
1.6050 to 1.6070 (20°C, 589nm)
500g
608-27-5
100.0
C6H5Cl2N
MFCD00007657
2,3 dichloraniline, 2,3-dichloranilin, 2,3-dichloro-phenylamine, 2,3-dichlorobenzenamine, 2,3-dichlorophenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro, aniline, 2,3-dichloro-7ci,8ci, benzenamine, 2,3-dichloro, unii-2bl4f1dxvn
C1=CC(=C(C(=C1)Cl)Cl)N
162.013
CHEBI:46636
Liquid After Melting
1.3700g/mL
Glass bottle
Brown
99%
98.5
99%
Cl2C6H3NH2
12, 621
BRPSAOUFIJSKOT-UHFFFAOYSA-N
2,3-dichloroaniline
11844
162.02
≥98.5%

Chemical Identifiers

608-27-5
162.013
BRPSAOUFIJSKOT-UHFFFAOYSA-N
11844
2,3-dichloroaniline
C6H5Cl2N
MFCD00007657
2,3 dichloraniline, 2,3-dichloranilin, 2,3-dichloro-phenylamine, 2,3-dichlorobenzenamine, 2,3-dichlorophenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro, aniline, 2,3-dichloro-7ci,8ci, benzenamine, 2,3-dichloro, unii-2bl4f1dxvn
CHEBI:46636
C1=CC(=C(C(=C1)Cl)Cl)N
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 210-157-9

RTECSNumber : CX9862625

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS Documents