Please login to your online account to display your discounted pricing

2,3-Dichloroaniline 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Dichloroaniline
Name Note 99%
CAS 608-27-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Formula Weight 162.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113095000

  SDS
thermo fisher scientific
113095000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid After Melting
Assay ≥98.5%
Formula Weight 162.02
Quantity 500g
Color Light brown to dark brown
Beilstein 12, 621
TSCA TSCA
RTECS Number CX9862625
EINECS Number 210-157-9
Density 1.3700g/mL
CAS Max % 100.0
Storage Information Product darkens on storage
CAS Min % 98.5
Refractive Index 1.6050 to 1.6070 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H5Cl2N
Chemical Name or Material 2, 3-Dichloroaniline
CAS 608-27-5
MDL Number MFCD00007657
Name Note 99%
Linear Formula Cl2C6H3NH2
Assay Percent Range 99%
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.