Learn More
2,3-Dichloroaniline, 99%, Thermo Scientific Chemicals
$339.27 - $339.27
Chemical Identifiers
CAS | 608-27-5 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.013 |
MDL Number | MFCD00007657 |
InChI Key | BRPSAOUFIJSKOT-UHFFFAOYSA-N |
Synonym | benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine |
PubChem CID | 11844 |
ChEBI | CHEBI:46636 |
IUPAC Name | 2,3-dichloroaniline |
SMILES | C1=CC(=C(C(=C1)Cl)Cl)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1152436
|
Thermo Scientific Chemicals
A1152436 |
500 g |
Each for $339.27
|
|
|||||
Description
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.
Solubility
Soluble in ether, alcohol and acetone. Slightly soluble in benzene. Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Chemical Identifiers
608-27-5 | |
162.013 | |
BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
11844 | |
2,3-dichloroaniline |
C6H5Cl2N | |
MFCD00007657 | |
benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
CHEBI:46636 | |
C1=CC(=C(C(=C1)Cl)Cl)N |
Specifications
608-27-5 | |
1.37 | |
115°C (239°F) | |
1.606 | |
500 g | |
472027 | |
Soluble in ether,alcohol and acetone. Slightly soluble in benzene. Insoluble in water. | |
C1=CC(=C(C(=C1)Cl)Cl)N | |
162.013 | |
CHEBI:46636 | |
99% |
20°C to 24°C | |
250°C to 252°C | |
C6H5Cl2N | |
MFCD00007657 | |
UN3442 | |
benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine | |
BRPSAOUFIJSKOT-UHFFFAOYSA-N | |
2,3-dichloroaniline | |
11844 | |
162.02 | |
2,3-Dichloroaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber : 210-157-9
RTECSNumber : CX9862625
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only