Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3'-Dichloroacetophenone, 98%

$204.92 - $875.43

Chemical Identifiers

Chemical Name or Material 2',3'-Dichloroacetophenone
CAS 56041-57-7
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00052988
Beilstein 2438815
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1266706

  SDS
alfa aesar™
A1266706
5g Each for $204.92

AAA1266714

  SDS
alfa aesar™
A1266714
25g Each for $875.43
Description & Specifications

Specifications

Formula Weight 189.04
CAS 56041-57-7
Assay 98%
Chemical Name or Material 2',3'-Dichloroacetophenone
Beilstein 2438815
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 106° to 108°C (2mmHg)
Molecular Formula C8H6Cl2O
MDL Number MFCD00052988
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.