Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloro-6-fluorophenylacetic acid, 97%

$70.19 - $167.78

Chemical Identifiers

Chemical Name or Material 2,3-Dichloro-6-fluorophenylacetic acid
CAS 886497-57-0
Molecular Formula C8H5Cl2FO2
Formula Weight 223.03
MDL Number MFCD04115875
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2635703

 
alfa aesar™
H2635703
1g Each for $167.78

AAH26357MD

 
alfa aesar™
H26357MD
250mg Each for $70.19
Description & Specifications

Specifications

Formula Weight 223.03
Chemical Name or Material 2,3-Dichloro-6-fluorophenylacetic acid
Assay 97%
Molecular Formula C8H5Cl2FO2
MDL Number MFCD04115875
CAS 886497-57-0
Melting Point 131° to 135°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.