Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloro-6-fluorobenzoic acid, 97%

$64.04 - $581.61

Chemical Identifiers

Chemical Name or Material 2,3-Dichloro-6-fluorobenzoic acid
CAS 32890-91-8
Molecular Formula C7H3Cl2FO2
Formula Weight 209.01
MDL Number MFCD00090506
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2628606

  SDS
alfa aesar™
H2628606
5g Each for $189.85

AAH2628603

  SDS
alfa aesar™
H2628603
1g Each for $64.04

AAH2628614

  SDS
alfa aesar™
H2628614
25g Each for $581.61
Description & Specifications

Specifications

Formula Weight 209.01
Health Hazard 1 Warning
Chemical Name or Material 2,3-Dichloro-6-fluorobenzoic acid
Assay 97%
Melting Point 134° to 136°C
Molecular Formula C7H3Cl2FO2
Quantity 5g
CAS 32890-91-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00090506
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.