missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloro-6-fluorobenzaldehyde, 97%

$91.11 - $807.80

Chemical Identifiers

CAS 95399-95-4
Molecular Formula C7H3Cl2FO
Formula Weight 193.01
MDL Number MFCD04115870
Chemical Name or Material 2,3-Dichloro-6-fluorobenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2656203 SDS Alfa Aesar™
H2656203
1g Each for $91.11
AAH2656206 SDS Alfa Aesar™
H2656206
5g Each for $269.27
AAH2656214 SDS Alfa Aesar™
H2656214
25g Each for $807.80
Specifications

Description

Applications
Used as solvents. T2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

95399-95-4
193.01
2,3-Dichloro-6-fluorobenzaldehyde
C7H3Cl2FO
MFCD04115870

Specifications

1g
C7H3Cl2FO
MFCD04115870
2,3-Dichloro-6-fluorobenzaldehyde
95399-95-4
193.01
56° to 60°C
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning