Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloro-6-fluoroanisole, 97%

Chemical Identifiers

Chemical Name or Material 2,3-Dichloro-6-fluoroanisole
Molecular Formula C7H5Cl2FO
Formula Weight 195.02
MDL Number MFCD09832260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3201606

  SDS
alfa aesar™
H3201606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3201603

  SDS
alfa aesar™
H3201603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,3-Dichloro-6-fluoroanisole
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Molecular Formula C7H5Cl2FO
Formula Weight 195.02
MDL Number MFCD09832260
Melting Point 47° to 50°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.