missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 241810250

 View more versions of this product

Catalog No. AC241810250



Specifications

Specifications

2, 3-Dichloro-5-(trifluoromethyl)pyridine
79°C
Glass bottle
1.549
Colorless to Yellow
25g
96.0
96% min. (GC)
MFCD00042243
ABNQGNFVSFKJGI-UHFFFAOYSA-N
2,3-dichloro-5-(trifluoromethyl)pyridine
112234
Liquid
1.5490g/mL
Authentic
1.4745 to 1.4765
176.0°C
8.0°C to 9.0°C
69045-84-7
100.0
C6H2Cl2F3N
2,3,5-dctf, 2,3-dichloro-5-trifluoro methylpyridine, 2,3-dichloro-5-trifluoromethyl pyridine, 2,3-dichloro-5-trifluoromethyl-pyridine, 5f2c8lx8ie, pubchem9237, pyridine, 2,3-dichloro-5-trifluoromethyl, unii-5f2c8lx8ie
C1=C(C=NC(=C1Cl)Cl)C(F)(F)F
216
216
97%

Chemical Identifiers

69045-84-7
216
ABNQGNFVSFKJGI-UHFFFAOYSA-N
112234
C1=C(C=NC(=C1Cl)Cl)C(F)(F)F
C6H2Cl2F3N
MFCD00042243
2,3,5-dctf, 2,3-dichloro-5-trifluoro methylpyridine, 2,3-dichloro-5-trifluoromethyl pyridine, 2,3-dichloro-5-trifluoromethyl-pyridine, 5f2c8lx8ie, pubchem9237, pyridine, 2,3-dichloro-5-trifluoromethyl, unii-5f2c8lx8ie
2,3-dichloro-5-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

SDS Documents