missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dichloro-5-fluoropyridine, 98%

$28.84 - $337.73

Chemical Identifiers

CAS 185985-40-4
Molecular Formula C5H2Cl2FN
Molecular Weight (g/mol) 165.976
MDL Number MFCD08669666
InChI Key YIYIQENCPIISLL-UHFFFAOYSA-N
Synonym pyridine,2,3-dichloro-5-fluoro, pyridine, 2,3-dichloro-5-fluoro, abbypharma ap-31-5779, 2,3-dichloro-5-fluoro-pyridine
PubChem CID 18520008
IUPAC Name 2,3-dichloro-5-fluoropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64373MD View Documents Alfa Aesar™
H64373MD
250mg Each for $28.84
 
AAH6437303 View Documents Alfa Aesar™
H6437303
1g Each for $85.14
 
AAH6437306 View Documents Alfa Aesar™
H6437306
5g Each for $337.73
 
Description

Description

Applications
2,3-Dichloro-5-fluoropyridine is a reagent that is used in the synthesis of 3-Chloro-5-fluoro Clodinafop Propargyl (C368645), which is an impurity of Clodinafop Propargyl.

Solubility
Slightly soluble in chloroform, methanol and water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

185985-40-4
165.976
YIYIQENCPIISLL-UHFFFAOYSA-N
18520008
C1=C(C=NC(=C1Cl)Cl)F
C5H2Cl2FN
MFCD08669666
pyridine,2,3-dichloro-5-fluoro, pyridine, 2,3-dichloro-5-fluoro, abbypharma ap-31-5779, 2,3-dichloro-5-fluoro-pyridine
2,3-dichloro-5-fluoropyridine

Specifications

250mg
C5H2Cl2FN
13831996
YIYIQENCPIISLL-UHFFFAOYSA-N
2,3-dichloro-5-fluoropyridine
18520008
Solid
2,3-Dichloro-5-fluoropyridine
185985-40-4
MFCD08669666
pyridine,2,3-dichloro-5-fluoro, pyridine, 2,3-dichloro-5-fluoro, abbypharma ap-31-5779, 2,3-dichloro-5-fluoro-pyridine
C1=C(C=NC(=C1Cl)Cl)F
165.976
165.98
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning