missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113305000
Description
Oxidation reagentSpecifications
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone | |
0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum) | |
0.2% max. (600°C) | |
210.0°C to 217.0°C | |
84-58-2 | |
100.0 | |
C8Cl2N2O2 | |
01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120 | |
1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile, ddq, dichlorodicyano-p-benzoquinone, dichlorodicyanobenzoquinone, dichlorodicyanoquinone | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile | |
6775 | |
Crystalline Powder or Crystals |
Authentic | |
Glass bottle | |
Yellow to Orange | |
500g | |
97.5 | |
97.5% min. (Iodometry) | |
10,902 | |
15,3072 | |
Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N | |
227.01 | |
227.01 | |
98% |
Chemical Identifiers
84-58-2 | |
227.01 | |
1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile, ddq, dichlorodicyano-p-benzoquinone, dichlorodicyanobenzoquinone, dichlorodicyanoquinone | |
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile |
C8Cl2N2O2 | |
HZNVUJQVZSTENZ-UHFFFAOYSA-N | |
6775 | |
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
GHS Signal Word: Danger
EINECSNumber : 201-542-2