Learn More
2,3-Dichloro-1-propanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 365090250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 3-Dichloro-1-propanol | |
96.0 | |
1.3500g/mL | |
93°C | |
96% min. (GC) | |
C3H6Cl2O | |
MFCD00040446 | |
1.35 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ether,acetone,ethanol | |
C(C(CCl)Cl)O | |
128.99 | |
128.99 |
616-23-9 | |
100.0 | |
182°C | |
Authentic | |
Glass bottle | |
1.4835 to 1.4855 | |
25g | |
2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954 | |
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N | |
2,3-dichloropropan-1-ol | |
12018 | |
97% |
Chemical Identifiers
616-23-9 | |
128.99 | |
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N | |
12018 | |
C(C(CCl)Cl)O |
C3H6Cl2O | |
MFCD00040446 | |
2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954 | |
2,3-dichloropropan-1-ol |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Suspected of causing genetic defects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 210-470-
RUO â Research Use Only