missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dibromothiophene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 258760050

 View more versions of this product

Catalog No. AC258760050



Specifications

Specifications

2, 3-Dibromothiophene
51°C
Glass bottle
2.2
Brown or Khaki-Orange to Yellow
97%
96.0
96% min. (GC)
MFCD00005418
ATRJNSFQBYKFSM-UHFFFAOYSA-N
2,3-dibromothiophene
76590
Powder and/or Chunks
2.2000g/mL
Authentic
1.6300 to 1.632
130°C
5g
3140-93-0
100.0
C4H2Br2S
2 3-dibromothiophene, 2,3-dibromo thiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, acmc-1ckid, dibromothiophene, ksc491i5b, pubchem5511, thiophene, 2,3-dibromo, thiophene,3-dibromo
C1=CSC(=C1Br)Br
241.92
241.92
97%

Chemical Identifiers

3140-93-0
241.92
ATRJNSFQBYKFSM-UHFFFAOYSA-N
76590
C1=CSC(=C1Br)Br
C4H2Br2S
MFCD00005418
2 3-dibromothiophene, 2,3-dibromo thiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, acmc-1ckid, dibromothiophene, ksc491i5b, pubchem5511, thiophene, 2,3-dibromo, thiophene,3-dibromo
2,3-dibromothiophene
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 221-542-6

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS Documents