Please login to your online account to display your discounted pricing

2,3-Dibromothiophene, 97%, ACROS Organics™

$88.20 - $88.20

Chemical Identifiers

Chemical Name or Material 2, 3-Dibromothiophene
Name Note 97%
CAS 3140-93-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C4H2Br2S
Formula Weight 241.92
MDL Number MFCD00005418
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC258760050

  SDS
thermo fisher scientific
258760050
Glass bottle 5g Each for $88.20
Description & Specifications

Specifications

Assay ≥96.0% (GC)
Color Yellow to orange, golden-khaki or brown
Formula Weight 241.92
Physical Form Powder and/or Chunks
Quantity 5g
TSCA TSCA
EINECS Number 221-542-6
Density 2.2000g/mL
CAS Max % 100.0
CAS Min % 96.0
Refractive Index 1.6310 to 1.6330 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Storage Information Product darkens on storage
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00005418
Assay Percent Range 97%
Name Note 97%
Molecular Formula C4H2Br2S
Chemical Name or Material 2, 3-Dibromothiophene
CAS 3140-93-0