missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromosuccinic acid, 98%

Manufacturer:  Alfa Aesar™ H6030614

 View more versions of this product

Catalog No. AAH6030614



Specifications

Specifications

2,3-Dibromosuccinic acid
25g
C4H4Br2O4
MFCD00004209
2,3-dibromo succinic acid
C(C(C(=O)O)Br)(C(=O)O)Br
275.88
275.88
98%
274°C
526-78-3
HOOCCHBrCHBrCOOH
UN3261
FJWGRXKOBIVTFA-UHFFFAOYSA-N
2,3-dibromobutanedioic acid
95324
Solid

Chemical Identifiers

526-78-3
275.88
FJWGRXKOBIVTFA-UHFFFAOYSA-N
95324
C(C(C(=O)O)Br)(C(=O)O)Br
C4H4Br2O4
MFCD00004209
2,3-dibromo succinic acid
2,3-dibromobutanedioic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents