missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromobutane, (+/-) + meso, 98+%

Manufacturer:  Alfa Aesar™ A1021906

 View more versions of this product

Catalog No. AAA1021906



Specifications

Specifications

2,3-Dibromobutane
166°C to 167°C
(±) + meso
C4H8Br2
1718916
BXXWFOGWXLJPPA-UHFFFAOYSA-N
2,3-dibromobutane
21508
≥98%
123°C (253°F)
5g
5408-86-6
MFCD00000146
butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
CC(C(C)Br)Br
215.916
215.93

Chemical Identifiers

5408-86-6
215.916
BXXWFOGWXLJPPA-UHFFFAOYSA-N
21508
CC(C(C)Br)Br
C4H8Br2
MFCD00000146
butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
2,3-dibromobutane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents