missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dibromobutane, (+/-) + meso, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1021922

 View more versions of this product

Catalog No. AAA1021922



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3-Dibromobutane
5408-86-6
166°C to 167°C
C4H8Br2
MFCD00000146
1718916
BXXWFOGWXLJPPA-UHFFFAOYSA-N
2,3-dibromobutane
21508
≥98%
(±) + meso
1.76
123°C (253°F)
1.5126
100 g
butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
CC(C(C)Br)Br
215.916
215.93

Chemical Identifiers

5408-86-6
215.916
BXXWFOGWXLJPPA-UHFFFAOYSA-N
21508
CC(C(C)Br)Br
C4H8Br2
MFCD00000146
butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
2,3-dibromobutane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 226-476-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only