missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3'-Dibromoacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1521014

 View more versions of this product

Catalog No. AAA1521014



Description

Description

Applications
It is employed as an intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

2,3'-Dibromoacetophenone
49°C to 52°C
18523-22-3
MFCD00082698
775786
MZBXSQBQPJWECM-UHFFFAOYSA-N
2-bromo-1-(3-bromophenyl)ethanone
519585
97%
Pungent
25g
C8H6Br2O
UN3261
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2,3'-dibromoacetophenone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
C1=CC(=CC(=C1)Br)C(=O)CBr
277.943
277.95

Chemical Identifiers

18523-22-3
277.943
MZBXSQBQPJWECM-UHFFFAOYSA-N
519585
C1=CC(=CC(=C1)Br)C(=O)CBr
C8H6Br2O
MFCD00082698
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2,3'-dibromoacetophenone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
2-bromo-1-(3-bromophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents