missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromo-6-chloropyridine, 95%

$260.14 - $766.57

Chemical Identifiers

CAS 885952-16-9
Molecular Formula C5H2Br2ClN
Formula Weight 271.34
MDL Number MFCD06798229
Chemical Name or Material 2,3-Dibromo-6-chloropyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2713603 SDS Alfa Aesar™
H2713603
1g Each for $766.57
AAH27136MD SDS Alfa Aesar™
H27136MD
250mg Each for $260.14
Specifications Description & Specifications

Chemical Identifiers

885952-16-9
271.34
2,3-Dibromo-6-chloropyridine
C5H2Br2ClN
MFCD06798229

Specifications

885952-16-9
1g
271.34
2,3-Dibromo-6-chloropyridine
69° to 71°C
C5H2Br2ClN
MFCD06798229
95%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger