Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromo-5-(trifluoromethyl)pyridine, 95%

Chemical Identifiers

Chemical Name or Material 2,3-Dibromo-5-(trifluoromethyl)pyridine
CAS 79623-38-4
Formula Weight 304.89
MDL Number MFCD10687233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6140703

 
alfa aesar™
H6140703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6140706

 
alfa aesar™
H6140706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61407MD

 
alfa aesar™
H61407MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 304.89
Chemical Name or Material 2,3-Dibromo-5-(trifluoromethyl)pyridine
Physical Form Solid
Assay 95%
MDL Number MFCD10687233
CAS 79623-38-4
Quantity 1g