Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromo-1-propene, tech. 85%, stab. with copper

$77.22 - $209.34

Chemical Identifiers

Chemical Name or Material 2,3-Dibromo-1-propene
CAS 513-31-5
Name Note Stabilized with copper
Purity Grade Technical Grade
Molecular Formula C3H4Br2
Formula Weight 199.88g/mol
MDL Number MFCD00000211
Beilstein 878169
Merck Index 14,3025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1209914

 
alfa aesar™
A12099-14
25g Each for $77.22

AAA1209922

 
alfa aesar™
A12099-22
100g Each for $209.34
Description & Specifications

Specifications

Boiling Point 141°C
Quantity 25g
Flash Point 81°C (177°F)
Chemical Name or Material 2,3-Dibromo-1-propene
Name Note Stabilized with copper
CAS 513-31-5
Purity Grade Technical Grade
Assay 85%
Molecular Formula C3H4Br2
Formula Weight 199.88g/mol
MDL Number MFCD00000211
Beilstein 878169
Merck Index 14,3025
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H341-H335-H227-H315
Causes serious eye damage.
Suspected of causing genetic defects.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protectio
Recommended Storage 2°C to 8°C
RTECS Number UC8200000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protectio

GHS H Statement
H318-H341-H335-H227-H315
Causes serious eye damage.
Suspected of causing genetic defects.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

2°C to 8°C