Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromo-1,4-butanediol, 98%

$88.36 - $213.96

Chemical Identifiers

Chemical Name or Material 2,3-Dibromo-1,4-butanediol
CAS 20163-90-0
Molecular Formula C4H8Br2O2
Formula Weight 247.93
MDL Number MFCD00011549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2526014

 
alfa aesar™
B2526014
25g Each for $88.36

AAB2526022

 
alfa aesar™
B2526022
100g Each for $213.96
Description & Specifications

Specifications

Formula Weight 247.93
Chemical Name or Material 2,3-Dibromo-1,4-butanediol
Melting Point 83° to 87°C
Boiling Point 148° to 150°C (1.5mmHg)
Assay 98%
Quantity 25g
Molecular Formula C4H8Br2O2
MDL Number MFCD00011549
CAS 20163-90-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.