Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Dibromo-1,1,3,3-tetrafluoropropene, 97%

$54.85 - $162.73

Chemical Identifiers

Chemical Name or Material 2,3-Dibromo-1,1,3,3-tetrafluoropropene
CAS 666-40-0
Molecular Formula CF2=CBrCF2Br
Formula Weight 271.83
MDL Number MFCD00236663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2155004

  SDS
alfa aesar™
B2155004
2g Each for $54.85

AAB2155009

  SDS
alfa aesar™
B2155009
10g Each for $162.73
Description & Specifications

Specifications

Formula Weight 271.83
Chemical Name or Material 2,3-Dibromo-1,1,3,3-tetrafluoropropene
TSCA TSCA
Assay 97%
Quantity 2g
Molecular Formula CF2=CBrCF2Br
MDL Number MFCD00236663
CAS 666-40-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.