missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diaminotoluene, 98%

Manufacturer:  Alfa Aesar™ A1564714

 View more versions of this product

Catalog No. AAA1564714



Specifications

Specifications

2,3-Diaminotoluene
56°C to 60°C
2687-25-4
MFCD00011589
AXNUJYHFQHQZBE-UHFFFAOYSA-N
3-methylbenzene-1,2-diamine
17593
98%
225°C
25g
C7H10N2
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
CC1=C(C(=CC=C1)N)N
122.171
122.17

Chemical Identifiers

2687-25-4
122.171
AXNUJYHFQHQZBE-UHFFFAOYSA-N
17593
CC1=C(C(=CC=C1)N)N
C7H10N2
MFCD00011589
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
3-methylbenzene-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents