Learn More
2,3-Diaminotoluene, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 305670050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 3-Diaminotoluene | |
2687-25-4 | |
100.0 | |
96% min. (HPLC) | |
C7H10N2 | |
MFCD00011589 | |
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine | |
AXNUJYHFQHQZBE-UHFFFAOYSA-N | |
3-methylbenzene-1,2-diamine | |
17593 | |
97% |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
CH3C6H3(NH2)2 | |
5g | |
Solubility in water: miscible. Other solubilities: soluble in diethyl ether and ethanol | |
CC1=CC=CC(N)=C1N | |
122.17 | |
122.17 |
Chemical Identifiers
2687-25-4 | |
122.17 | |
AXNUJYHFQHQZBE-UHFFFAOYSA-N | |
17593 | |
CC1=CC=CC(N)=C1N |
C7H10N2 | |
MFCD00011589 | |
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine | |
3-methylbenzene-1,2-diamine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause cancer.<b
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 220-248-5
RTECSNumber : XS9550000
TSCA : TSCA
RUO â Research Use Only