missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Diaminonaphthalene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC159590250

 View more versions of this product

Catalog No. AC159590250



Specifications

Specifications

2,3-Diaminonaphthalene
>110°C
Glass bottle
Brown to Brown-Green
25g
96% min. (HPLC)
C10H6(NH2)2
2,3 diaminonapthalene
XTBLDMQMUSHDEN-UHFFFAOYSA-N
naphthalene-2,3-diamine
69872
Crystalline Powder
1.0968g/mL
Authentic
1.0968
193.0°C to 199.0°C
771-97-1,91-59-8
C10H10N2
13,207
Solubility in water: < 0.1% (20°C).
C1=CC=C2C=C(C(=CC2=C1)N)N
158.2
158.2
97%

Chemical Identifiers

771-97-1
158.2
2,3 diaminonapthalene
naphthalene-2,3-diamine
C10H10N2
XTBLDMQMUSHDEN-UHFFFAOYSA-N
69872
C1=CC=C2C=C(C(=CC2=C1)N)N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 212-241-

SDS Documents