Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diamino-6-methoxypyridine dihydrochloride, 96%

$62.68 - $525.86

Chemical Identifiers

Chemical Name or Material 2,3-Diamino-6-methoxypyridine dihydrochloride
CAS 94166-62-8
Molecular Formula C6H9N3O·2HCl
Formula Weight 212.08
MDL Number MFCD07787253
Synonym 6-Methoxy-2,3-pyridinediamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2647203

  SDS
alfa aesar™
H2647203
1g Each for $149.18

AAH2647206

  SDS
alfa aesar™
H2647206
5g Each for $525.86

AAH26472MD

  SDS
alfa aesar™
H26472MD
250mg Each for $62.68
Description & Specifications

Specifications

Formula Weight 212.08
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD07787253
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 6-Methoxy-2,3-pyridinediamine hydrochloride
Molecular Formula C6H9N3O·2HCl
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
Chemical Name or Material 2,3-Diamino-6-methoxypyridine dihydrochloride
Melting Point 167° to 171°C
CAS 94166-62-8
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.