Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Diamino-5-chloropyridine, 98+%

Chemical Identifiers

Chemical Name or Material 2,3-Diamino-5-chloropyridine
CAS 25710-20-7
Molecular Formula C5H6ClN3
Formula Weight 143.58
MDL Number MFCD01230986
UN Number UN2811
Beilstein 119434
Synonym 5-Chloro-2,3-diaminopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1952606

 
alfa aesar™
L1952606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1952614

 
alfa aesar™
L1952614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.58
Assay ≥98%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 170° to 174°C
Quantity 5g
Chemical Name or Material 2,3-Diamino-5-chloropyridine
MDL Number MFCD01230986
CAS 25710-20-7
Beilstein 119434
Synonym 5-Chloro-2,3-diaminopyridine
Molecular Formula C5H6ClN3
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.