missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-5-bromopyridine, 97%

CAS: 38875-53-5 | C5H7BrN3 | 189.04 g/mol

Supplier:  Thermo Scientific Chemicals L1943514

 View more versions of this product

Catalog No. AAL1943514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3-Diamino-5-bromopyridine
153°C to 161°C
MFCD00460094
UN2811
2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine
NC1=CC(Br)=C[NH+]=C1N
189.04
188.03
38875-53-5
C5H7BrN3
25 g
119436
YRGMYJUKFJPNPD-UHFFFAOYSA-O
2,3-diamino-5-bromopyridin-1-ium
691156
97%

Chemical Identifiers

38875-53-5
189.04
YRGMYJUKFJPNPD-UHFFFAOYSA-O
691156
NC1=CC(Br)=C[NH+]=C1N
C5H7BrN3
MFCD00460094
2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine
2,3-diamino-5-bromopyridin-1-ium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 254-172-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only