Please login to your online account to display your discounted pricing

2,3-Diamino-5-bromopyridine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3-Diamino-5-bromopyridine
CAS 38875-53-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H6BrN3
Formula Weight 188.03
MDL Number MFCD00460094
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350330250

  SDS
fisher scientific acros
35033-0250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350330050

  SDS
thermo fisher scientific
35033-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder or Needles
Formula Weight 188.03
Quantity 25g
Color Light yellow to purple or light brown
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 3-Diamino-5-bromopyridine
Packaging Glass bottle
MDL Number MFCD00460094
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C5H6BrN3
CAS 38875-53-5
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.