Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3-Cyclopentenopyridine, 99%

$67.95 - $210.95

Chemical Identifiers

Chemical Name or Material 2,3-Cyclopentenopyridine
CAS 533-37-9
Molecular Formula C8H9N
Formula Weight 119.17
MDL Number MFCD00005933
Synonym 7-Azaindane; 6,7-Dihydro-5H-cyclopenta[b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2589606

  SDS
alfa aesar™
H2589606
5g Each for $67.95

AAH2589614

  SDS
alfa aesar™
H2589614
25g Each for $210.95
Description & Specifications

Specifications

Formula Weight 119.17
Molecular Formula C8H9N
MDL Number MFCD00005933
Synonym 7-Azaindane; 6,7-Dihydro-5H-cyclopenta[b]pyridine
Boiling Point 87° to 88°C (11mmHg)
Health Hazard 1 Warning
Flash Point 67°C (153°F)
Quantity 5g
CAS 533-37-9
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material 2,3-Cyclopentenopyridine
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.