missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3-Chlorophenyl)ethylamine, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 13078-79-0 |
---|---|
Molecular Formula | C8H10ClN |
Molecular Weight (g/mol) | 155.63 |
MDL Number | MFCD00047957 |
InChI Key | NRHVNPYOTNGECT-UHFFFAOYSA-N |
Synonym | 2-3-chlorophenyl ethylamine, 2-3-chlorophenyl ethanamine, 3-chlorophenethylamine, 2-3-chlorophenyl ethan-1-amine, 2-3-chloro-phenyl-ethylamine, 3-chlorophenylethanamine, 3'-chlorophenethylamine, m-chlorophenethylamine, 2-m-chlorophenyl ethylamine, benzeneethanamine, 3-chloro |
PubChem CID | 139381 |
IUPAC Name | 2-(3-chlorophenyl)ethanamine |
SMILES | C1=CC(=CC(=C1)Cl)CCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC342160050
|
Thermo Scientific Chemicals
342160050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC342160250
|
Thermo Scientific Chemicals
342160250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13078-79-0 | |
155.63 | |
NRHVNPYOTNGECT-UHFFFAOYSA-N | |
139381 | |
C1=CC(=CC(=C1)Cl)CCN |
C8H10ClN | |
MFCD00047957 | |
2-3-chlorophenyl ethylamine, 2-3-chlorophenyl ethanamine, 3-chlorophenethylamine, 2-3-chlorophenyl ethan-1-amine, 2-3-chloro-phenyl-ethylamine, 3-chlorophenylethanamine, 3'-chlorophenethylamine, m-chlorophenethylamine, 2-m-chlorophenyl ethylamine, benzeneethanamine, 3-chloro | |
2-(3-chlorophenyl)ethanamine |
Specifications
13078-79-0 | |
100.0 | |
111.0°C to 113.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5480 to 1.5500 | |
MFCD00047957 | |
1.119 | |
NRHVNPYOTNGECT-UHFFFAOYSA-N | |
2-(3-chlorophenyl)ethanamine | |
139381 | |
98% |
97.5 | |
1.1190g/mL | |
>109°C | |
98% | |
C8H10ClN | |
H2N(CH2)2C6H4Cl | |
5g | |
2-3-chlorophenyl ethylamine, 2-3-chlorophenyl ethanamine, 3-chlorophenethylamine, 2-3-chlorophenyl ethan-1-amine, 2-3-chloro-phenyl-ethylamine, 3-chlorophenylethanamine, 3'-chlorophenethylamine, m-chlorophenethylamine, 2-m-chlorophenyl ethylamine, benzeneethanamine, 3-chloro | |
C1=CC(=CC(=C1)Cl)CCN | |
155.63 | |
155.63 | |
2-(3-Chlorophenyl)ethylamine, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W
GHS Signal Word: Warning
TSCA : TSCA
RUO â Research Use Only