missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Chlorophenyl)benzimidazole, 95%

$124.14 - $1,480.43

Chemical Identifiers

CAS 22868-35-5
Molecular Formula C13H9ClN2
Molecular Weight (g/mol) 228.679
MDL Number MFCD01438282
InChI Key BGYHTIIVIGGCIF-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
PubChem CID 767074
IUPAC Name 2-(3-chlorophenyl)-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64245MD View Documents Alfa Aesar™
H64245MD
250mg Each for $124.14
 
AAH6424503 View Documents Alfa Aesar™
H6424503
1g Each for $370.11
 
AAH6424506 View Documents Alfa Aesar™
H6424506
5g Each for $1,480.43
 
Specifications

Chemical Identifiers

22868-35-5
228.679
BGYHTIIVIGGCIF-UHFFFAOYSA-N
767074
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)Cl
C13H9ClN2
MFCD01438282
2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
2-(3-chlorophenyl)-1H-benzimidazole

Specifications

250mg
C13H9ClN2
2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)Cl
228.679
228.68
95%
22868-35-5
MFCD01438282
BGYHTIIVIGGCIF-UHFFFAOYSA-N
2-(3-chlorophenyl)-1H-benzimidazole
767074
Solid
2-(3-Chlorophenyl)benzimidazole