missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Chlorophenyl)benzimidazole, 95%

Manufacturer:  Alfa Aesar™ H6424506

 View more versions of this product

Catalog No. AAH6424506



Specifications

Specifications

2-(3-Chlorophenyl)benzimidazole
22868-35-5
MFCD01438282
BGYHTIIVIGGCIF-UHFFFAOYSA-N
2-(3-chlorophenyl)-1H-benzimidazole
767074
Solid
5g
C13H9ClN2
2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)Cl
228.679
228.68
95%

Chemical Identifiers

22868-35-5
228.679
BGYHTIIVIGGCIF-UHFFFAOYSA-N
767074
C1=CC=C2C(=C1)NC(=N2)C3=CC(=CC=C3)Cl
C13H9ClN2
MFCD01438282
2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
2-(3-chlorophenyl)-1H-benzimidazole
SDS Documents