Please login to your online account to display your discounted pricing

2-(3-Chlorophenoxy)ethanethioamide, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
CAS 35370-95-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C8H8ClNOS
Linear Formula ClC6H4OCH2CSNH2
Formula Weight 201.68
MDL Number MFCD00052479
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB01327EA

  SDS
fisher scientific company
SPB01327EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.68
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
Assay Percent Range 95%+
Molecular Formula C8H8ClNOS
Linear Formula ClC6H4OCH2CSNH2
Packaging Amber Glass Bottle
MDL Number MFCD00052479
CAS 35370-95-7