Please login to your online account to display your discounted pricing

2-(3-Chlorophenoxy)ethanethioamide, ≥95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB01327EA

 View more versions of this product

Catalog No. SPB01327EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 201.68
Quantity 10g
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Linear Formula ClC6H4OCH2CSNH2
Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
Molecular Formula C8H8ClNOS
Assay Percent Range 95%+
MDL Number MFCD00052479
CAS 35370-95-7

Specifications

Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
CAS 35370-95-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C8H8ClNOS
Linear Formula ClC6H4OCH2CSNH2
Formula Weight 201.68
MDL Number MFCD00052479