missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3'-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine 97.0+%, TCI America™
Supplier: TCI America C31631G
Specifications
2-(3′-Chlorobiphenyl-4-yl)-4,6-diphenyl-1,3,5-triazine | |
206°C | |
C27H18ClN3 | |
PGIGQFHBEHJSQN-UHFFFAOYSA-N | |
2-[4-(3-chlorophenyl)phenyl]-4,6-diphenyl-1,3,5-triazine | |
91972125 | |
≥97.0% (HPLC,N) |
1443049-84-0 | |
White-Yellow | |
1 g | |
C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=C(C=C3)C4=CC(=CC=C4)Cl)C5=CC=CC=C5 | |
419.912 | |
419.91 | |
Crystalline Powder |
Chemical Identifiers
1443049-84-0 | |
419.912 | |
91972125 | |
C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=C(C=C3)C4=CC(=CC=C4)Cl)C5=CC=CC=C5 |
C27H18ClN3 | |
PGIGQFHBEHJSQN-UHFFFAOYSA-N | |
2-[4-(3-chlorophenyl)phenyl]-4,6-diphenyl-1,3,5-triazine |
Safety and Handling
TSCA : No