Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Chloro-4-fluorophenyl)indole, 98%

$147.22 - $147.22

Chemical Identifiers

Chemical Name or Material 2-(3-Chloro-4-fluorophenyl)indole
CAS 1868-88-8
Molecular Formula C14H9ClFN
Formula Weight 245.69
MDL Number MFCD00068179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1292003

  SDS
alfa aesar™
A1292003
1g Each for $147.22
Description & Specifications

Specifications

Formula Weight 245.69
Chemical Name or Material 2-(3-Chloro-4-fluorophenyl)indole
Assay 98%
Molecular Formula C14H9ClFN
MDL Number MFCD00068179
CAS 1868-88-8
Melting Point 170° to 173°C
Quantity 1g