Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Chloro-4-fluorophenyl)ethanol, 96%

$50.80 - $169.32

Chemical Identifiers

Chemical Name or Material 2-(3-Chloro-4-fluorophenyl)ethanol
CAS 340825-21-0
Molecular Formula C8H8ClFO
Formula Weight 174.6
MDL Number MFCD06201121
Synonym 3-Chloro-4-fluorophenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3229103

 
alfa aesar™
H3229103
1g Each for $169.32

AAH32291MD

 
alfa aesar™
H32291MD
250mg Each for $50.80
Description & Specifications

Specifications

Formula Weight 174.6
Chemical Name or Material 2-(3-Chloro-4-fluorophenyl)ethanol
Assay 96%
Molecular Formula C8H8ClFO
MDL Number MFCD06201121
CAS 340825-21-0
Synonym 3-Chloro-4-fluorophenethyl alcohol
Quantity 1g