Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Butynyloxy)tetrahydropyran, 98%

$94.81 - $94.81

Chemical Identifiers

Chemical Name or Material 2-(3-Butynyloxy)tetrahydropyran
CAS 40365-61-5
Molecular Formula C9H14O2
Formula Weight 154.21
MDL Number MFCD00012352
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5338506

 
alfa aesar™
H5338506
5g Each for $94.81
Description & Specifications

Specifications

Formula Weight 154.21
Chemical Name or Material 2-(3-Butynyloxy)tetrahydropyran
Assay 98%
Molecular Formula C9H14O2
MDL Number MFCD00012352
CAS 40365-61-5
Health Hazard 1 Danger
Boiling Point 92° to 95°C (18mmHg)
Health Hazard 2 GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Flash Point 73°C (163°F)
Health Hazard 3 GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-(3-Butynyloxy)tetrahydropyran is used as a reagent in the synthesis of Phomoidride D, a bridged, polycyclic, natural compound that has the ability to inhibit squalene synthase and can be used as an antihypercholesterolemic agent. It is also used as a reagent to prepare Rugulactone, a natural compound that is isolated from Cryptocarya rugulosa.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.