missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3-Bromophenyl)propan-2-ol, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444620050
Specifications
2-(3-Bromophenyl)propan-2-ol | |
5g | |
96% min. (GC) | |
2-3-bromo-phenyl-propan-2-ol, 2-3-bromophenyl propan-2-ol, 2-3-bromophenyl-2-propanol, 2-3-bromophenyl-propan-2-ol, 3-alpha-hydroxyisopropyl-1-bromobenzene, acmc-209hjh | |
CC(C)(C1=CC(=CC=C1)Br)O | |
215.09 | |
215.09 |
100°C (3.0 Torr) | |
30951-66-7 | |
C9H11BrO | |
ZRFMJMFYMQAUDO-UHFFFAOYSA-N | |
2-(3-bromophenyl)propan-2-ol | |
15072383 | |
97% |
Chemical Identifiers
30951-66-7 | |
215.09 | |
2-3-bromo-phenyl-propan-2-ol, 2-3-bromophenyl propan-2-ol, 2-3-bromophenyl-2-propanol, 2-3-bromophenyl-propan-2-ol, 3-alpha-hydroxyisopropyl-1-bromobenzene, acmc-209hjh | |
2-(3-bromophenyl)propan-2-ol |
C9H11BrO | |
ZRFMJMFYMQAUDO-UHFFFAOYSA-N | |
15072383 | |
CC(C)(C1=CC(=CC=C1)Br)O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning