Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Bromo-4-methoxyphenyl)ethylamine, 98%

Chemical Identifiers

Chemical Name or Material 2-(3-Bromo-4-methoxyphenyl)ethylamine
CAS 159465-27-7
Molecular Formula C9H12BrNO
Formula Weight 230.11
MDL Number MFCD00153891
UN Number UN2735
Beilstein 3250744
Synonym 3-Bromo-4-methoxyphenethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987403

  SDS
alfa aesar™
L1987403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1987406

  SDS
alfa aesar™
L1987406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.11
Chemical Name or Material 2-(3-Bromo-4-methoxyphenyl)ethylamine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C9H12BrNO
Assay 98%
Synonym 3-Bromo-4-methoxyphenethylamine
CAS 159465-27-7
Boiling Point 114°C (1mmHg)
MDL Number MFCD00153891
Beilstein 3250744
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Flash Point >110°C (230°F)
UN Number UN2735
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.