missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Benzofuran, 99.50%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC105480500

 View more versions of this product

Catalog No. AC105480500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.4% (GC)
118.13
Yellow to green to yellow-brown
17, 54
100.0
2, 3-Benzofuran
99.4
0.1% max. (K.F.)
Authentic
99.5%
Crystalline Powder or Crystals
50mL
1.0700g/mL
15, 1090
Coumarone
MFCD00005847
Glass bottle
1.5655 to 1.5685 (20°C, 589nm)
271-89-6
C8H6O

Chemical Identifiers

271-89-6
100.0
C8H6O
MFCD00005847
2, 3-Benzofuran
99.4
99.5%
118.13
Coumarone
≥99.4% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Suspected of causing cancer.
Flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning