missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,6-Trifluorotoluene, 98%, Thermo Scientific™
$40.08 - $424.49
Chemical Identifiers
CAS | 119916-25-5 |
---|---|
Molecular Formula | C7H5Cl3 |
Molecular Weight (g/mol) | 195.467 |
MDL Number | MFCD12922564 |
InChI Key | UZYYBZNZSSNYSA-UHFFFAOYSA-N |
Synonym | 2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference |
PubChem CID | 16378 |
IUPAC Name | 1,2,4-trichloro-3-methylbenzene |
SMILES | CC1=C(C=CC(=C1Cl)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63897MD
|
Thermo Scientific Chemicals
H63897MD |
250 mg |
Each for $40.08
|
|
|||||
AAH6389703
|
Thermo Scientific Chemicals
H6389703 |
1 g |
Each for $109.86
|
|
|||||
AAH6389706
|
Thermo Scientific Chemicals
H6389706 |
5 g |
Each for $424.49
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
119916-25-5 | |
195.467 | |
UZYYBZNZSSNYSA-UHFFFAOYSA-N | |
16378 | |
CC1=C(C=CC(=C1Cl)Cl)Cl |
C7H5Cl3 | |
MFCD12922564 | |
2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference | |
1,2,4-trichloro-3-methylbenzene |
Specifications
119916-25-5 | |
250 mg | |
C7H5Cl3 | |
2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference | |
CC1=C(C=CC(=C1Cl)Cl)Cl | |
195.467 | |
146.11 | |
2,3,6-Trifluorotoluene |
Colorless to Yellow | |
Liquid | |
MFCD12922564 | |
UZYYBZNZSSNYSA-UHFFFAOYSA-N | |
1,2,4-trichloro-3-methylbenzene | |
16378 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures