missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',6'-Trifluoropropiophenone, 97+%

$65.22 - $239.35

Chemical Identifiers

CAS 243666-18-4
Molecular Formula C9H7F3O
Formula Weight 188.15
MDL Number MFCD00061222
Chemical Name or Material 2',3',6'-Trifluoropropiophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2443403 SDS Alfa Aesar™
B2443403
1g Each for $65.22
AAB2443406 SDS Alfa Aesar™
B2443406
5g Each for $239.35
Specifications Description & Specifications

Chemical Identifiers

243666-18-4
188.15
2',3',6'-Trifluoropropiophenone
C9H7F3O
MFCD00061222

Specifications

1g
C9H7F3O
MFCD00061222
≥97%
243666-18-4
188.15
2',3',6'-Trifluoropropiophenone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning