Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluorobenzyl bromide, 98%

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluorobenzyl bromide
CAS 151412-02-1
Molecular Formula C7H4BrF3
Formula Weight 225.01
MDL Number MFCD00061208
UN Number UN3265
Beilstein 8762828
Synonym alpha-Bromo-2,3,6-trifluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1990903

 
alfa aesar™
A1990903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1990906

 
alfa aesar™
A1990906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.01
Chemical Name or Material 2,3,6-Trifluorobenzyl bromide
Assay 98%
Molecular Formula C7H4BrF3
MDL Number MFCD00061208
CAS 151412-02-1
UN Number UN3265
Beilstein 8762828
Synonym alpha-Bromo-2,3,6-trifluorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 90°C (194°F)
Melting Point 114°C (dec.)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.