Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluorobenzyl alcohol, 97%

$53.04 - $53.04

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluorobenzyl alcohol
CAS 114152-19-1
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00061205
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2005603

  SDS
alfa aesar™
B2005603
1g Each for $53.04
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 2,3,6-Trifluorobenzyl alcohol
Health Hazard 1 Warning
CAS 114152-19-1
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00061205
Assay 97%
Molecular Formula C7H5F3O
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Flash Point >110°C (230°F)
Melting Point 44° to 48°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.