missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,6-Trifluorobenzoyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC312760010

 View more versions of this product

Catalog No. AC312760010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 3, 6-Trifluorobenzoyl chloride
Clear slightly yellow
189807-20-3
100.0
Glass bottle
Liquid
98%
F3C6HCOCl
MFCD00061203
1.48g/mL
1g
97.5
Authentic
1.4815 to 1.4835 (20°C, 589nm)
≥97.5%
C7H2ClF3O
194.54

Chemical Identifiers

189807-20-3
100.0
C7H2ClF3O
194.54
2, 3, 6-Trifluorobenzoyl chloride
97.5
98%
F3C6HCOCl
MFCD00061203
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger